missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LGI2 Polyclonal antibody specifically detects LGI2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | LGI2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ10675, glioma inactivated 2, KIAA1916MGC126810, leucine-rich repeat LGI family, member 2, LGI1-like protein 2, MGC126808 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 10 amino acid synthetic peptide from near the center of human LGI2. The immunogen is located within amino acids 290 - 340 of LGI2. Amino Acid Squence: IYKYDESWTK |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?