missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LGI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118395
This item is not returnable.
View return policy
Description
LGI1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The leucine-rich, glioma inactivated gene 1 (LGI1) was first identified as a candidate tumor suppressor gene for glioma and may play a role in other cancers. LGI1 is a member of a family of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 serves as a ligand to ADAM22, a metalloprotease localized at the synapse. Mutations in LGI1 account for nearly half of autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Two isoforms of LGI1 are known to exist.
Specifications
| LGI1 | |
| Polyclonal | |
| Unconjugated | |
| LGI1 | |
| ADLTE; ADPAEF; ADPEAF; BB130740; EPITEMPIN; Epitempin 1; Epitempin-1; EPT; ETL1; IB1099; leucine rich glioma inactivated 1; leucine-rich glioma inactivated 1; leucine-rich glioma-inactivated protein 1; leucine-rich repeat LGI family, member 1; leucine-rich, glioma inactivated 1; LGI1; OTTHUMP00000020121; OTTHUMP00000020122; UNQ775/PRO1569 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 252892, 9211 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O95970, Q8K4Y5 | |
| LGI1 | |
| Synthetic peptide within rat LGI1 aa 200-250. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction