missing translation for 'onlineSavingsMsg'
Learn More

LGD-6972, MedChemExpress

Product Code. 16495906
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495906 100 mg 100mg
16481447 50 mg 50mg
16471269 1 mg 1mg
16465238 10 mg 10mg
16457957 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495906 Supplier Medchem Express Supplier No. HY12525
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress LGD-6972 is a selective and orally active glucagon receptor antagonist. LGD-6972 has the potential for type 2 diabetes research.

LGD-6972 is a selective and orally active glucagon receptor antagonist. LGD-6972 has the potential for type 2 diabetes research.

Specifications

Chemical Name or Material LGD-6972
CAS 1207989-09-0
Color White
Purity Grade Notes Research
Molecular Formula C43H46N2O5S
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (177.83 mM; Need ultrasonic)
SMILES O=C(NCCS(=O)(O)=O)C1=CC=C(C[C@H](C2=CC=C(C3=CC=C(C(C)(C)C)C=C3)C=C2)C(NC4=CC=C(C5=C(C)C=C(C)C=C5C)C=C4)=O)C=C1
Molecular Weight (g/mol) 702.9
Formula Weight 702.9
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.