missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALSL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561143
This item is not returnable.
View return policy
Description
Immunogen sequence: LNNSLSSPVQ ADVYFPRLIV PFCGHIKGGM RPGKKVLVMG IVDLNPESFA ISLTCGDSED PPADVAIELK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Does not bind lactose, and may not bind carbohydrates.
Specifications
| LGALSL | |
| Polyclonal | |
| Unconjugated | |
| LGALSL | |
| 1110067D22Rik; A530071M23; galectin like; Galectin-like protein; Galectin-related protein; galectin-related protein A; GRP; Grpa; HSPC159; Lectin galactoside-binding-like protein; lectin galactoside-binding-like protein A; lectin, galactoside binding like; lectin, galactoside binding-like; lectin, galactoside-binding-like; Lgalsl; Lgalsla; RGD1307414 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29094 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3ZCW2 | |
| LGALSL | |
| Recombinant protein corresponding to Human LGALSL | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction