missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LGALSL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590100
This item is not returnable.
View return policy
Description
Immunogen sequence: MAGSVADSDA VVKLDDGHLN NSLSSPVQAD VYFPRLIVPF CGHIKGGMRP GKKVLVMGIV DLNPESFAIS LTCGDSEDPP ADVAIELKAV FTDRQLLRNS CISGERGEEQ SAIPYFPFIP DQPFRVEILC EHPRFRVFVD GHQLFDFYHR IQTLSAIDTI KINGDLQITK LG; Positive Samples: Mouse brain.
Does not bind lactose, and may not bind carbohydrates.
Specifications
| LGALSL | |
| Polyclonal | |
| Unconjugated | |
| LGALSL | |
| 1110067D22Rik; A530071M23; galectin like; Galectin-like protein; Galectin-related protein; galectin-related protein A; GRP; Grpa; HSPC159; Lectin galactoside-binding-like protein; lectin galactoside-binding-like protein A; lectin, galactoside binding like; lectin, galactoside binding-like; lectin, galactoside-binding-like; Lgalsl; Lgalsla; RGD1307414 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216551 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.36 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8VED9 | |
| LGALSL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human LGALSL (NP_0549002). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction