missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LGALS3BP (Human) Recombinant Protein
Human LGALS3BP full-length ORF ( AAH02403.1, 1 a.a. - 585 a.a.) recombinant protein with GST-tag at N-terminal.
Specifications
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|---|---|
| Gene ID (Entrez) | 3959 |
Description
- Sequence: MTPPRLFWVWLLVAGTQGVNDGDMRLADGGATNQGRVEIFYRGQWGTVCDNLWDLTDASVVCRALGFENATQALGRAAFGQGSGPIMLDEVQCTGTEASLADCKSLGWLKSNCRHERDAGVVCTNETRSTHTLDLSRELSEALGQIFDSQRGCDLSISVNVQGEDALGFCGHTVILTANLEAQALWKEPGSNVTMSVDAECVPMVRDLLRYFYSRRIDITLSSVKCFHKLASAYGARQLQGYCASLFAILLPQDPSFQMPLDLYAYAVATGDALLEKLCLQFLAWNFEALTQAEAWPSVPTDLLQLLLPRSDLAVPSELALLKAVDTWSWGERASHEEVEGLVEKIRFPMMLPEELFELQFNLSLYWSHEALFQKKTLQALEFHTVPFQLLARYKGLNLTEDTYKPRIYTSPTWSAFVTDSSWSARKSQLVYQSRRGPLVKYSSDYFQAPSDYRYYPYQSFQTPQHPSFLFQDKRVSWSLVYLPTIQSCWNYGFSCSSDELPVLGLTKSGGSDRTIAYENKALMLCEGLFVADVTDFEGWKAATPSALDTNSSKSTSSFPCPAGHFNGFRTVIRPFYLTNSSGVD
Specifications
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| 3959 |
Safety and Handling
missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title