missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LG 100268
GREENER_CHOICE

Product Code. 18714579
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18714579 25 mg 25mg
18726278 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18714579 Supplier Tocris Bioscience™ Supplier No. 5920/25
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective RXR agonist; also activates RXR/PPARγ heterodimers

LG 100268 is a potent and selective retinoid X receptor (RXR) agonist (EC50 = 4 nM). Displays >1000-fold selectivity for RXR over RAR. Activates RXR homodimers to induce transcriptional activation. Induces tissue transglutaminase (TGase) activity in HL-60 human promyelocytic leukemia cell line. Also activates RXR/PPARγ heterodimers.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)cyclopropyl]-3-pyridinecarboxylic acid
CAS 153559-76-3
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C24H29NO2
Formula Weight Observed MW: 363.49
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.