missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LFNG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584802
This item is not returnable.
View return policy
Description
Immunogen sequence: TKKFHRARLD LLLETWISRH KEMTFIFTDG EDEALARHTG NVVITNCSAA H Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 88%.
LFNG belongs to evolutionarily conserved glycosyltransferases that act in the Notch signaling pathway to define boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta-1,3-N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling. This protein is predicted to be a single-pass type II Golgi membrane protein but it may also be secreted and proteolytically processed like the related proteins in mouse and Drosophila (PMID: 9187150).
Specifications
| LFNG | |
| Polyclonal | |
| Unconjugated | |
| LFNG | |
| AW061165; beta-1,3-N-acetylglucosaminyltransferase lunatic fringe; LFNG; LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; lunatic fringe; lunatic fringe gene homolog; MGC128664 protein; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; SCDO3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3955 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NES3 | |
| LFNG | |
| Recombinant protein corresponding to Human LFNG. Recombinant protein control fragment (Product #RP-103673). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction