missing translation for 'onlineSavingsMsg'
Learn More

Lexibulin, MedChemExpress

Product Code. 16444008
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444008

Brand: Medchem Express HY10498

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lexibulin (CYT-997) is a potent and orally active tubulin polymerisation inhibitor with IC50s of 10-100 nM in cancer cell lines; with potent cytotoxic and vascular disrupting activity in vitro and in vivo. Lexibulin induces cell apoptosis and induces mitochondrial ROS generation in GC cells.

Lexibulin (CYT-997) is a potent and orally active tubulin polymerisation inhibitor with IC50s of 10-100 nM in cancer cell lines

Specifications

Chemical Name or Material Lexibulin
CAS 917111-44-5
Color White
Purity Grade Notes Research
Molecular Formula C24H30N6O2
Quantity 10 mg
Synonym CYT-997
Solubility Information DMSO : ≥ 100 mg/mL (230.13 mM)
SMILES CCC[C@H](NC1=C(C)C=NC(C2=CC=C(NC(NCC)=O)C(OC)=C2)=N1)C3=CN=CC=C3
Molecular Weight (g/mol) 434.53
Formula Weight 434.53
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.