missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewis Y, Rabbit anti-Human, Clone: BR96, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81077-0.025MG
This item is not returnable.
View return policy
Description
Lewis Y Monoclonal specifically detects Lewis Y in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Lewis Y | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| The antibody binds Lewis Y with a Ka of 0.1 uM. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| ELISA, Immunohistochemistry | |
| BR96 | |
| Unconjugated | |
| PBS | |
| H3396 cultured line established from a metastasis of a human breast adenocarcinoma. | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction