missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewis Y, Mouse anti-Human, Clone: BR96, Novus Biologicals™
Mouse Monoclonal Antibody
£225.00 - £463.00
Specifications
| Antigen | Lewis Y |
|---|---|
| Clone | BR96 |
| Concentration | 1 mg/mL |
| Dilution | ELISA, Immunohistochemistry |
| Applications | ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18397640
|
Novus Biologicals
NBP2-81076-0.025MG |
0.025 mg |
£225.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18288079
|
Novus Biologicals
NBP2-81076-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lewis Y Monoclonal specifically detects Lewis Y in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Lewis Y | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| H3396 cultured line established from a metastasis of a human breast adenocarcinoma. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| BR96 | |
| ELISA, Immunohistochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| IgG1 κ | |
| Protein A purified | |
| The antibody binds Lewis Y with a Ka of 0.1 uM. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title