missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lewis Y Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Lewis Y Humanized Biosimilar
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283123
GREENER_CHOICE
View Documents
Invitrogen™
MA558691
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283125
View Documents
Invitrogen™
MA558692
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term MB 311.

The Lewis Y (Le^y) gene is not actually a single gene but refers to a carbohydrate antigen known as Lewis Y or Le^y antigen, which is part of the Lewis blood group system. This antigen is synthesized by the fucosylation of precursor substances, which involves enzymes encoded by the FUT1 (fucosyltransferase 1) and FUT2 genes. These genes are responsible for the addition of fucose, a hexose deoxy sugar, to the precursor oligosaccharides. The expression of Lewis Y antigen has been extensively studied in the context of cancer biology as it is commonly found on the surface of epithelial and endothelial cells but overexpressed in various types of cancers, including colorectal, gastric, and ovarian carcinomas. The overexpression and altered glycosylation patterns of Le^y antigen in tumors have made it a potential biomarker for cancer diagnosis and a target for immunotherapeutic approaches.
Specifications

Specifications

Lewis Y Humanized Biosimilar
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.