missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewis Y Antibody (A70-A/A9), Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £402.00
Specifications
| Antigen | Lewis Y |
|---|---|
| Clone | A70-A/A9 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunofluorescence 1-2ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658606
|
Novus Biologicals
NBP2-50073SS |
0.02 mg |
£232.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606378
|
Novus Biologicals
NBP2-50073-0.1mg |
0.1 mg |
£402.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
| Lewis Y | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Tumor Biomarkers | |
| 10 mM PBS with 0.05% BSA | |
| Live Ls174T cells (human colon carcinoma cell line) | |
| Primary | |
| Store at 4°C. |
| A70-A/A9 | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunofluorescence 1-2ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title