missing translation for 'onlineSavingsMsg'
Learn More

Lewis A Blood Group Antigen Antibody (HE-193), Novus Biologicals™

Product Code. 18394262 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394262 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394262 Supplier Novus Biologicals Supplier No. NBP309164

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lewis A Blood Group Antigen Monoclonal specifically detects Lewis A Blood Group Antigen in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lewis A Blood Group Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone HE-193
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1 ug/ml
Formulation PBS
Host Species Mouse
Immunogen Mixture of erythrocytes of group A1 and glycoprotein fraction isolated from saliva of secretors with blood group A.
Purification Method Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.