missing translation for 'onlineSavingsMsg'
Learn More

Lewis A,B Blood Group Antigens Antibody (HEA164), Biotin, Novus Biologicals™

Product Code. 18354402 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354402 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354402 Supplier Novus Biologicals Supplier No. NBP312056B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lewis A,B Blood Group Antigens Monoclonal antibody specifically detects Lewis A,B Blood Group Antigens in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lewis A,B Blood Group Antigens
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone HEA164
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias Le-a, Le-b, Lewis A, Lewis A Blood Group Antigen, Lewis B, lewis B blood group antigen
Host Species Rabbit
Immunogen This antibody was raised by immunising mice with human carcinoma cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.