missing translation for 'onlineSavingsMsg'
Learn More

Levulinic Acid, TRC

Product Code. 17974975
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974975 500 g 500g
17970525 50 g 50g
17932325 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974975 Supplier Toronto Research Chemicals Supplier No. TRCL379500500G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-76-2
Molecular Formula C5 H8 O3
Molecular Weight (g/mol) 116.12
Synonym Levulinic Acid, Pentanoic acid, 4-oxo-, Levulinic acid (8CI), 4-Oxopentanoic acid, 3-Acetylpropanoic acid, 3-Acetylpropionic acid, 3-Oxobutanecarboxylic acid, 4-Ketovaleric acid, 4-Oxovaleric acid, Laevulinic acid, Levulic acid, NSC 3716, β-Acetylpropionic acid, γ-Ketovaleric acid
IUPAC Name 4-oxopentanoic acid
SMILES CC(=O)CCC(=O)O

Specifications

Quantity 500 g
InChI Formula InChI=1S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)
Formula Weight 116.0473
Chemical Name or Material Levulinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.