missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levosulpiride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Levosulpiride |
| CAS | 23672-07-3 |
| Analyte or Component Names | Levosulpiride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H23 N3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 5-(Aminosulfonyl)-N-[[(2S)-1-ethyl-2-pyrrolidinyl]methyl]-2-methoxybenzamide, Benzamide, 5-(aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxy-, (S)-, o-Anisamide, N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-sulfamoyl-, (-)- (8CI), (-)-Sulpiride, L-Sulpiride, Lesuride, Levobren, Levopraid, Levosulpiride, S-(-)-Sulpiride, S-Sulpiride, l-Sulpiride, levopride, sulpid |
| InChI Formula | InChI=1S/C15H23N3O4S/c1-3-18-8-4-5-11(18)10-17-15(19)13-9-12(23(16,20)21)6-7-14(13)22-2/h6-7,9,11H,3-5,8,10H2,1-2H3,(H,17,19)(H2,16,20,21)/t11-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCN1CCC[C@H]1CNC(=O)c2cc(ccc2OC)S(=O)(=O)N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction