missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Levonorgestrel, 98%

Product Code. 15567049 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15567049

Brand: Thermo Scientific Chemicals 461100010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 797-63-7
Melting Point 232.0°C to 239.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C21H28O2
Quantity 1 g
Synonym levonorgestrel, norgestrel, d-norgestrel, --norgestrel, levonova, microval, postinor, mirena, plan b, jadelle
InChI Key WWYNJERNGUHSAO-XUDSTZEESA-N
SMILES CCC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34
IUPAC Name (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 312.45
PubChem CID 13109
ChEBI CHEBI:6443
Formula Weight 312.45
Percent Purity 98%
Chemical Name or Material Levonorgestrel
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Levonorgestrel
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H351-Suspected of causing cancer.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.