missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levomepromazine Hydrochloride, Mikromol™
Description
Levomepromazine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Levomepromazine hydrochloride |
| CAS | 1236-99-3 |
| Analyte or Component Names | Levomepromazine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 N2 O S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Levomepromazine Hydrochloride, (betaR)-2-Methoxy-N,N,beta-trimethyl-10H-phenothiazine-10-propanamine monohydrochloride, (R)-2-Methoxy-N,N,beta-trimethyl-10H-phenothiazine-10-propanamine monohydrochloride, (-)-10-[3-(Dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine monohydrochloride, (-)-Methotrimeprazine hydrochloride, Levomepromazine hydrochloride, Levomepromazinium chloride, Levopromazine hydrochloride, Methotrimeprazine hydrochloride, Neurocil |
| InChI Formula | InChI=1S/C19H24N2OS.ClH/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21;/h5-11,14H,12-13H2,1-4H3;1H/t14-;/m1./s1 |
| SMILES | Cl.COc1ccc2Sc3ccccc3N(C[C@H](C)CN(C)C)c2c1 |
| IUPAC Name | (2R)-3-(2-methoxyphenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?