missing translation for 'onlineSavingsMsg'
Learn More

(±)-Levomepromazine-d6, MedChemExpress

Product Code. 16461819
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461819 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461819 Supplier Medchem Express Supplier No. HY19489S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (±)-Levomepromazine D6 ((±)-Methotrimeprazine D6) is the deuterium labeled Methotrimeprazine, which is a D3 dopamine and Histamine H1 receptor antagonist.

(±)-Levomepromazine D6 ((±)-Methotrimeprazine D6) is the deuterium labeled Methotrimeprazine, which is a D3 dopamine and Histamine H1 receptor antagonist.

Specifications

Chemical Name or Material (±)-Levomepromazine-d6
CAS 1189805-51-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C19H18D6N2OS
Color Off-White
Synonym (±)-Methotrimeprazine-d6 dl-Methotrimeprazine-d6
SMILES C1C=C2SC3=CC=C(OC)C=C3N(CC(CN(C([2H])([2H])[2H])C([2H])([2H])[2H])C)C2=CC=1
Molecular Weight (g/mol) 334.51
Quantity 1 mg
Formula Weight 334.51
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.