missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin, 98%

Product Code. 15488509
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488509 5 g 5g
15478509 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488509 Supplier Thermo Scientific Alfa Aesar Supplier No. J66943.06
  • ONLINE EXCLUSIVE PRICE
    £46.41
    £37.65 / 5g
    Save £8.76
    18% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance, and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division. | CAS: 100986-85-4 | C18H20FN3O4 | 361.373 g/mol

Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

Solubility
Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100986-85-4
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
MDL Number MFCD00865049
InChI Key GSDSWSVVBLHKDQ-JTQLQIEISA-N
PubChem CID 149096
ChEBI CHEBI:63598
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Specifications

Type Levofloxacin
Physical Form Powder
Melting Point 218°C to 227°C
Packaging Glass Bottle
Quantity 5 g
Flash Point >110°C (230°F)
Sensitivity Light sensitive
Beilstein 7327015
Boiling Point 572°C
Solubility Information Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane
Formula Weight 361.37
Percent Purity 98%
Chemical Name or Material Levofloxacin
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Levofloxacin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Levofloxacin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.