missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levodopa, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Levodopa |
| CAS | 59-92-7 |
| Analyte or Component Names | Levodopa |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H11 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Levodopa |
| InChI Formula | InChI=1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1 |
| Solution Type | Neat |
| SMILES | N[C@@H](Cc1ccc(O)c(O)c1)C(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction