missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levobupivacaine hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Description
Levobupivacaine hydrochloride exerts anaesthetic and analgesic effects through reversible blockade of neuronal sodium channel. Levobupivacaine hydrochloride can inhibit impulse transmission and conduction in cardiovascular and other tissues, possessing certain cardiac and CNS toxicity. Levobupivacaine hydrochloride is metabolized by hepatic cytochrome P450 (CYP450) enzymes in vivo. Levobupivacaine hydrochloride can also induce ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer.
Specifications
Specifications
| Chemical Name or Material | Levobupivacaine hydrochloride |
| CAS | 27262-48-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H29ClN2O |
| Quantity | 200 mg |
| Synonym | (S)-(-)-Bupivacaine monohydrochloride |
| Solubility Information | DMSO : 100 mg/mL (307.80 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1N(CCCC)CCCC1)NC2=C(C)C=CC=C2C.[H]Cl |
| Molecular Weight (g/mol) | 324.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction