missing translation for 'onlineSavingsMsg'
Learn More

Levobunolol hydrochloride, MedChemExpress

Product Code. 16488648
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488648 10 mg 10mg
16454527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488648 Supplier Medchem Express Supplier No. HYB1035
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Levobunolol (l-Bunolol) hydrochloride is a potent and nonselective β-adrenergic receptor antagonist. Levobunolol hydrochloride is an ocular hypotensive agent and lowers mean intraocular pressure (IOP). Levobunolol hydrochloride can be used for glaucoma and superior oblique myokymia (SOM) research.

Levobunolol (l-Bunolol) hydrochloride is a potent and nonselective β-adrenergic receptor antagonist. Levobunolol hydrochloride is an ocular hypotensive agent and lowers mean intraocular pressure (IOP). Levobunolol hydrochloride can be used for glaucoma and superior oblique myokymia (SOM) research.

Specifications

Chemical Name or Material Levobunolol hydrochloride
CAS 27912-14-7
Color White
Purity Grade Notes Research
Molecular Formula C17H26ClNO3
Quantity 10 mg
Synonym l-Bunolol hydrochloride
Solubility Information DMSO : ≥ 62.5 mg/mL (190.64 mM) ∣H2O : 50 mg/mL (152.51 mM; Need ultrasonic)
SMILES O=C1CCCC2=C1C=CC=C2OC[C@@H](O)CNC(C)(C)C.[H]Cl
Molecular Weight (g/mol) 327.85
Formula Weight 327.85
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.