missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Levilactobacillus brevis ATCC 8287 Culti-Loop

Product Code. 12988988
Change view
Click to view available options
No. per Pack:
5 Pk.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
12988988 5 Pk. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.
Product Code. 12988988 Supplier Thermo Scientific™ Supplier No. R4603053
  • ONLINE EXCLUSIVE PRICE
    £301.00
    £238.25 / Pack of 5
    Save £62.75
    20% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.

Culti-Loops containing the quality control microorganism Levilactobacillus brevis ATCC 8287.

Thermo Scientific Oxoid Culti-Loops containing the quality control microorganism Levilactobacillus brevis ATCC 8287, previously Lactobacillus brevis ATCC 8287.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 8287
Certifications/Compliance 21 CFR Part 820
Format Preserved Gel Matrix Suspended in Inoculating Loop
Manufacturing Origin United States
Product Type Culti-Loop
No. per Pack 5 Pk.
Species Lactobacillus species
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.