missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | CCL6/C10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RM0183-7J13 |
| Conjugate | APC-Cyanine7 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Symbols | Ccl6 |
| Host Species | Rat |
| Immunogen | Mouse recombinant CCL6/C10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?