missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam, MedChemExpress

Product Code. 16472016
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472016 500 mg 500mg
16468096 100 mg 100mg
16434237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472016 Supplier Medchem Express Supplier No. HYB0106
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Levetiracetam, an antiepileptic agent, binds the synaptic vesicle protein SV2A. Levetiracetam enhances Temozolomide effect on glioblastoma stem cell proliferation and apoptosis. Levetiracetam modulates HDAC levels ultimately silencing MGMT, thus increasing Temozolomide effectiveness. A chemosensitizer agent.

Levetiracetam, an antiepileptic agent, binds the synaptic vesicle protein SV2A. Levetiracetam enhances Temozolomide effect on glioblastoma stem cell proliferation and apoptosis. Levetiracetam modulates HDAC levels ultimately silencing MGMT, thus increasing Temozolomide effectiveness. A chemosensitizer agent.

Specifications

Chemical Name or Material Levetiracetam
CAS 102767-28-2
Color White
Purity Grade Notes Research
Quantity 500 mg
Synonym UCB L059
Solubility Information H2O : ≥ 85 mg/mL (499.38 mM) ∣DMSO : ≥ 85 mg/mL (499.38 mM)
SMILES NC([C@@H](N(CCC1)C1=O)CC)=O
Molecular Weight (g/mol) 170.21
Percent Purity 99.21%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.