missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Levcromakalim

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Levcromakalim is an active enantiomer of the prototypical Kir6 (KATP) channel opener Cromakalim. Hypotensive and airways relaxant. IC50 = 490 nM in guinea pig trachea.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 286.33 |
| Purity | >99% |
| Molecular Weight (g/mol) | 286.331 |
| Chemical Name or Material | Levcromakalim |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction