missing translation for 'onlineSavingsMsg'
Learn More

Levamisole hydrochloride, MedChemExpress

Product Code. 16498467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498467 5 g 5g
16475798 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498467 Supplier Medchem Express Supplier No. HY13666
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Levamisole ((-)-Tetramisole) hydrochloride is an anthelmintic and immunomodulator belonging to a class of synthetic imidazothiazole derivatives. Levamisole hydrochloride has antiviral effects against HSV.

Levamisole ((-)-Tetramisole) hydrochloride is an anthelmintic and immunomodulator belonging to a class of synthetic imidazothiazole derivatives. Levamisole hydrochloride has antiviral effects against HSV.

Specifications

Chemical Name or Material Levamisole hydrochloride
Physical Form Solid
CAS 16595-80-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C11H13ClN2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym (-)-Tetramisole hydrochloride
Solubility Information H2O : 50 mg/mL (207.68 mM; Need ultrasonic) ∣DMSO : 15.5 mg/mL (64.38 mM; Need ultrasonic and warming) ∣Ethanol : 12.5 mg/mL (51.92 mM; Need ultrasonic)
SMILES [H]Cl.C12=N[C@@H](C3=CC=CC=C3)CN1CCS2
Molecular Weight (g/mol) 240.75
Formula Weight 240.75
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.