missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leupeptin Hemisulfate

Product Code. 15493809
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15493809

Brand: Thermo Scientific Chemicals J61188.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reversible protease inhibitor which inhibits cathepsin B, calpain and trypsin | CAS: 103476-89-7 | C20H39N6O4+

Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

Solubility
Soluble in water, ethanol, dimethyl formamide and acetic acid.

Notes
Store in a cool place. Incompatible with strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103476-89-7
Molecular Formula C20H39N6O4+

Specifications

Color White
Product Type Leupeptin
Formula Weight 475.59
Solubility Soluble in water at 50mg/ml
Content And Storage -30°C to -10°C
Form Powder
Quantity 100 mg
compliance-icons
Product Identifier
  • Leupeptin hemisulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Leupeptin hemisulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.