missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leupeptin hemisulfate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Leupeptin hemisulfate is a membrane-permeable thiol protease inhibitor that inhibits Cathepsin B, Cathepsin H and Cathepsin L, and also impairs amphisome-lysosome fusion. Leupeptin hemisulfate also exhibits anti-inflammatory effect.
Specifications
Specifications
| Chemical Name or Material | Leupeptin hemisulfate |
| Physical Form | Powder |
| CAS | 103476-89-7 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H38N6O4.1/2H2SO4 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : 83.33 mg/mL (175.21 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C[C@@H](NC([C@@H](NC([C@@H](NC(C)=O)CC(C)C)=O)CC(C)C)=O)CCCNC(N)=N.[0.5H2SO4] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction