missing translation for 'onlineSavingsMsg'
Learn More

Melford Leupeptin Hemisulfate

Product Code. 30018381
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018381 25 mg 25mg
30018379 5 mg 5mg
30018380 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018381 Supplier Melford Supplier No. L220350.025
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.

Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin. Working concentration: 1-10uM

Specifications

CAS 103476-89-7
Molecular Formula C20H38N6O4 1/2H2SO4
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.