missing translation for 'onlineSavingsMsg'
Learn More

Melford Leupeptin Hemisulfate

Product Code. 30018379
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. Quantity unitSize
30018379 5 mg 5mg
30018380 10 mg 10mg
30018381 25 mg 25mg
3 options
This item is not returnable. View return policy

Product Code. 30018379

Brand: Melford L220350.005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.

Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin. Working concentration: 1-10uM

Specifications

CAS 103476-89-7
Molecular Formula C20H38N6O4 1/2H2SO4
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.