missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene B4 R1 Rabbit anti-Mouse, Rat, Clone: 4R2X8, Novus Biologicals™
Rabbit Monoclonal Antibody
£159.00 - £396.00
Specifications
| Antigen | Leukotriene B4 R1 |
|---|---|
| Clone | 4R2X8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18381405
|
Novus Biologicals
NBP3-15458-20UL |
20 μg |
£159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18376963
|
Novus Biologicals
NBP3-15458-100UL |
100 μg |
£396.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Leukotriene B4 R1 Monoclonal antibody specifically detects Leukotriene B4 R1 in Mouse, Rat samples. It is validated for Western BlotEspecificaciones
| Leukotriene B4 R1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1241 | |
| IgG | |
| Affinity purified |
| 4R2X8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
| A synthetic peptide corresponding to a sequence within amino acids 253-352 of human Leukotriene B4 R1 (Q15722). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto