missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene B4 R1 Antibody (202/7B1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64831MFV610
This item is not returnable.
View return policy
Description
Leukotriene B4 R1 Monoclonal antibody specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry
Specifications
| Leukotriene B4 R1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 202/7B1 | |
| Flow Cytometry | |
| chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
| HeLa cell transfectants expressing human BLTR. | |
| 0.1 mL | |
| GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1241 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction