missing translation for 'onlineSavingsMsg'
Learn More

Leukotriene A4 Hydrolase/LTA4H Antibody, DyLight 680, Novus Biologicals™

Product Code. 18363053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363053 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363053 Supplier Novus Biologicals Supplier No. NBP299029FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Leukotriene A4 Hydrolase/LTA4H Polyclonal specifically detects Leukotriene A4 Hydrolase/LTA4H in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leukotriene A4 Hydrolase/LTA4H
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Leukotriene A4 Hydrolase/LTA4H (NP_000886.1; Met 1-Asp 611).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4048
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.