missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene A4 Hydrolase/LTA4H Antibody (6G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26161-100ul
This item is not returnable.
View return policy
Description
Leukotriene A4 Hydrolase/LTA4H Monoclonal antibody specifically detects Leukotriene A4 Hydrolase/LTA4H in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| Leukotriene A4 Hydrolase/LTA4H | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 6G7 | |
| Western Blot 1:500-1:5000, ELISA | |
| EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase | |
| A synthesized peptide derived from Human Leukotriene A4 Hydrolase/LTA4H [UniProt P09960] | |
| 100 μL | |
| Lipid and Metabolism | |
| 4048 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction