missing translation for 'onlineSavingsMsg'
Learn More

Leukadherin-1, MedChemExpress

Product Code. 16446886
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446886 100 mg 100mg
16429307 5 mg 5mg
16416758 10 mg 10mg
16412837 50 mg 50mg
16411196 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16446886 Supplier Medchem Express Supplier No. HY15701
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Leukadherin-1, a specific agonist of the leukocyte surface integrin CD11b/CD18, increases CD11b/CD18-dependent cell adhesion to fibrinogen with an EC50 of 4 μM. Leukadherin-1 enhances leukocyte adhesion to ligands (such as ICAM-1) and vascular endothelium and thus reduces leukocyte transendothelial migration and influx to the injury sites. Leukadherin-1 suppresses innate inflammatory signaling.

Leukadherin-1, a specific agonist of the leukocyte surface integrin CD11b/CD18, increases CD11b/CD18-dependent cell adhesion to fibrinogen with an EC50 of 4 μM. Leukadherin-1 enhances leukocyte adhesion to ligands (such as ICAM-1) and vascular endothelium and thus reduces leukocyte transendothelial migration and influx to the injury sites. Leukadherin-1 suppresses innate inflammatory signaling.

Specifications

Chemical Name or Material Leukadherin-1
CAS 344897-95-6
Color Orange
Purity Grade Notes Research
Molecular Formula C22H15NO4S2
Quantity 100 mg
Solubility Information DMSO : 5 mg/mL (11.86 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(C2=CC=C(/C=C(SC(N3CC4=CC=CC=C4)=S)/C3=O)O2)C=C1
Molecular Weight (g/mol) 421.49
Formula Weight 421.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.