missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Leucyl-cystinyl Aminopeptidase/LNPEP Antibody (E2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27948
This item is not returnable.
View return policy
Description
Leucyl-cystinyl Aminopeptidase/LNPEP Monoclonal antibody specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot
Specifications
| Leucyl-cystinyl Aminopeptidase/LNPEP | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) | |
| angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase | |
| Synthetic peptide derived from human Leucyl-cystinyl Aminopeptidase/LNPEP (YFAATQFEPLAAR) | |
| 100 μg | |
| Immunology | |
| 4012 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| E2 | |
| Western Blot 1:1000 - 1:5000 | |
| Q9UIQ6 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction