missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leucyl-cystinyl Aminopeptidase/LNPEP Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81854AF405
This item is not returnable.
View return policy
Description
Leucyl-cystinyl Aminopeptidase/LNPEP Polyclonal antibody specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Leucyl-cystinyl Aminopeptidase/LNPEP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 405 | |
| angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase | |
| LNPEP antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human LNPEP. The immunogen is located within amino acids 900 - 950 of LNPEP. Amino Acid Squence: SLNGDNFRTQKLSFI | |
| 0.1 mL | |
| Immunology | |
| 4012 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction