Learn More
Greiner Bio-One Leucosep™ Polypropylene Round Bottom Centrifugation Tubes
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
Brand:  Greiner Bio-One 227288_X
Product Code. 13007723
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Enrichment directly from whole blood
  • Simplified filling through porous barrier
  • Rapid separation in 15 minutes at room temperature
  • Removal of erythrocytes and granulocytes
  • No recontamination with erythrocytes
  • Service Plan
    No