Learn More
Greiner Bio-One Leucosep Centrifuge Tubes with Porous Barrier
GREENER_CHOICE
Product Code. 15805547
Change view
Click to view available options
Length (Metric):
115 mm
Bottom Shape:
Conical
Unit Size:
Pack of 25
Product Code. Length (Metric) Bottom Shape unitSize
15805547 115 mm Conical Pack of 25
1 options
Product Code. 15805547 Supplier Greiner Bio-One Supplier No. 227290P
Only null left
Add to basket
Edge
Add to basket
Efficient separation of lymphocytes and mononuclear cells.
  • Non-cytotoxic, non-pyrogenic
  • Enrichment directly from whole blood
  • Simplified filling through porous barrier
  • Rapid separation in 15 minutes at room temperature
  • No additional laboratory equipment required, asepticall
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Natural
Color Translucent; print of graduation: blue
Material Polypropylene
Sterility Sterile
Length (Metric) 115 mm
Bottom Shape Conical
Closure Included Yes
Graduated Yes
Product Type Centrifuge Tube