missing translation for 'onlineSavingsMsg'
Learn More

Leucomycins (>85%), TRC

Product Code. 30137363
Change view
Click to view available options
Quantity:
250 mg
5 g
10 g
Unit Size:
10g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137363 10 g 10g
30134171 5 g 5g
17905065 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137363 Supplier Toronto Research Chemicals Supplier No. TRCL33032010G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1392-21-8
Molecular Formula C35H59NO13
Synonym Ayermicina, C 637, Kitasamycin, Sineptina, Stereomycine, Syneptine;
SMILES C[C@@H]1C/C=C/C=C/[C@@H]([C@@H](C[C@@H]([C@@H]([C@H]([C@@H](CC(=O)O1)O)OC)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C)O[C@H]3C[C@@]([C@H]([C@@H](O3)C)O)(C)O)N(C)C)O)CC=O)C)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C35H59NO13/c1-19-16-23(14-15-37)31(32(44-8)25(39)17-26(40)45-20(2)12-10-9-11-13-24(19)38)49-34-29(41)28(36(6)7)30(21(3)47-34)48-27-18-35(5,43)33(42)22(4)46-27/h9-11,13,15,19-25,27-34,38-39,41-43H,12,14,16-18H2,1-8H3/b10-9+,13-11+/t19-,20-,21-,22+,23+,24+,25-,27+,28-,29-,30-,31+,32+,33+,34+,35-/m1/s1
Chemical Name or Material Leucomycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.