missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leucomalachite Green

Product Code. 11370378
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11370378 10 g 10g
11380378 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11370378 Supplier Thermo Scientific Chemicals Supplier No. 043758.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 129-73-7 | C23H26N2 | 330.475 g/mol

Leucomalachite Green is a reagent for the quantitative colorimetric micro determination of hemoglobin and other heme compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23H26N2
Molecular Weight (g/mol) 330.475
MDL Number MFCD00008315
InChI Key WZKXBGJNNCGHIC-UHFFFAOYSA-N
Synonym 4,4'-Benzylidenebis(N,N-dimethylaniline); Malachite green leuco base
PubChem CID 67215
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C

Specifications

Melting Point 100°C to 102°C
Beilstein 2140506
Merck Index 14,5699
Solubility Information Soluble in water (<0.1mg/ml),chloroform,benzene,ethyl ether,ethanol and toluene.
Formula Weight 330.47
Sensitivity Air Sensitive
Quantity 10 g
Physical Form Powder
Chemical Name or Material Leucomalachite Green
compliance-icons
Product Identifier
  • Leucomalachite green
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4,4'-(phenylmethylene)bis[N,N-dimethylbenzenamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.