missing translation for 'onlineSavingsMsg'
Learn More

Leucine (L-Leucine), Mikromol™

Product Code. 17907744
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17907744 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17907744 Supplier Mikromol Supplier No. MM1470.00
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Leucine (L-Leucine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Leucine
CAS 61-90-5
Analyte or Component Names L-Leucine
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O2
Certifications/Compliance ISO 17025
Synonym Phenylalanine Imp. A (EP), Leucine, (S)-2-amino-4-methylpentanoic acid, Leucine, L- (8CI), (2S)-2-Amino-4-methylpentanoic acid, (S)-(+)-Leucine, (S)-2-Amino-4-methylpentanoic acid, (S)-2-Amino-4-methylvaleric acid, (S)-Leucine, 100: PN: US20070066537 PAGE: 17 claimed protein, 124: PN: EP2071334 SEQID: 136 claimed protein, 124: PN: WO2009077864 SEQID: 136 claimed protein, 164: PN: WO2010045512 SEQID: 173 unclaimed protein, 325: PN: WO2010059829 SEQID: 353 unclaimed protein, 45: PN: WO2008005407 PAGE: 5 claimed protein, 8: PN: US20090069547 PAGE: 10 claimed protein, L-(+)-Leucine, L-2-Amino-4-methylpentanoic acid, L-Norvaline, 4-methyl-, L-alpha-Aminoisocaproic acid, Leu, NSC 46709, Pentanoic acid, 2-amino-4-methyl-, (S)-, Phenylalanine Impurity A
InChI Formula InChI=1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1
SMILES CC(C)C[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
Molecular Weight (g/mol) 131.17
Quantity 250 mg
Formula Weight 131.0946 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.