missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leu-Enkephalin

Product Code. 15464009
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464009 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15464009 Supplier Thermo Scientific Chemicals Supplier No. J61408.MB
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81678-16-2615.684 g/mol

Endogenous opioid neurotransmitter/neuromodulator that is an agonist at ? and δ opioid receptors, localization in brain parallels the localization of δ receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Endogenous opioid neurotransmittereuromodulator that is an agonist at μ and δ opioid receptors, localization in brain parallels the localization of δ receptors.

Solubility
Soluble in water (1 mg/ml ).
TRUSTED_SUSTAINABILITY

Specifications

CAS 81678-16-2
Color White
Content And Storage Store at -20°C
Format Lyophilized Powder
Molecular Weight (g/mol) 615.684
Product Type Leu-Enkephalin
Quantity 25 mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.