missing translation for 'onlineSavingsMsg'
Learn More

Leu-AMS R enantiomer, MedChemExpress

Product Code. 16441119
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16441119

Brand: Medchem Express HY108900A1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Leu-AMS R enantiomer is the R enatiomer of Leu-AMS. Leu-AMS is a potent inhibitor of leucyl-tRNA synthetase (LRS) and inhibits the growth of bacteria.

Leu-AMS R enantiomer is the R enatiomer of Leu-AMS. Leu-AMS is a potent inhibitor of leucyl-tRNA synthetase (LRS) and inhibits the growth of bacteria.

Specifications

Chemical Name or Material Leu-AMS R enantiomer
Color White
Purity Grade Notes Research
Molecular Formula C16H25N7O7S
Quantity 1 mg
Solubility Information DMSO : 50.5 mg/mL (109.91 mM; Need ultrasonic and warming)
SMILES O[C@@H]1[C@@H](COS(NC([C@H](N)CC(C)C)=O)(=O)=O)O[C@@H](N2C(N=CN=C3N)=C3N=C2)[C@@H]1O
Molecular Weight (g/mol) 459.48
Formula Weight 459.48
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.