missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leu-AMS R enantiomer, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Leu-AMS R enantiomer is the R enatiomer of Leu-AMS. Leu-AMS is a potent inhibitor of leucyl-tRNA synthetase (LRS) and inhibits the growth of bacteria.
Specifications
Specifications
| Chemical Name or Material | Leu-AMS R enantiomer |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H25N7O7S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 50.5 mg/mL (109.91 mM; Need ultrasonic and warming) |
| SMILES | O[C@@H]1[C@@H](COS(NC([C@H](N)CC(C)C)=O)(=O)=O)O[C@@H](N2C(N=CN=C3N)=C3N=C2)[C@@H]1O |
| Molecular Weight (g/mol) | 459.48 |
| Formula Weight | 459.48 |
| Grade | Research |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction