missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Letrozole
Description
Potent, reversible non-steroidal aromatase inhibitor (IC50 = 11.5 nM). Displays antitumor effects in several animal models. Suppresses the endogenous aromatase-induced proliferation of MCF-7 cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 285.3 |
| Purity | >99% |
| Molecular Weight (g/mol) | 285.31 |
| Chemical Name or Material | Letrozole |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?