missing translation for 'onlineSavingsMsg'
Learn More

Letrozole, MedChemExpress

Product Code. p-200096330
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16461116

Brand: Medchem Express HY14248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Letrozole (CGS 20267) is a potent, selective, reversible and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. Letrozole selective inhibits estrogen biosynthesis, and can be used for the research of breast cancer.

Letrozole (CGS 20267) is a potent, selective, reversible and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. Letrozole selective inhibits estrogen biosynthesis, and can be used for the research of breast cancer.

Specifications

Chemical Name or Material Letrozole
Physical Form Powder
CAS 112809-51-5
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C17H11N5
For Use With (Application) Cancer-programmed cell death
Synonym CGS 20267
Solubility Information DMSO : 50 mg/mL (175.25 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C(N2C=NC=N2)C3=CC=C(C#N)C=C3)C=C1
Molecular Weight (g/mol) 285.3
Formula Weight 285.3
Percent Purity 99.79%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.